3-Kloori-4-fluorifenoli ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

A14382.06 A14382.14
A14382.06EA 63.9 EUR
A14382.06 A14382.14
3-Kloori-4-fluorifenoli ≥97%
3-Kloori-4-fluorifenoli

Purity: 97% CAS No.: 2613-23-2 Molecular Formula: C6H4ClFO Formula Weight: 146.55 Flash Point: 108°(226°F) Odour: Phenol-like Boiling Point: 103-104°/11mm Beilstein Reference: 3236443 UN No.: 3261 MDL No.: MFCD00002257

Kaava: C₆H₄ClFO
Kiehumispiste: 103…104 °C (11 mmHg)
Sulamispiste: 38…43 °C
Leimahduspiste: 108 °C (226 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002257
CAS-numero: 2613-23-2
EINECS: 220-042-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR