D(+)-biotiini ≥98%

Toimittaja: Thermo Fisher Scientific

A14207.06 A14207.03 A14207.09 A14207.14
A14207.06EA 537 EUR
A14207.06 A14207.03 A14207.09 A14207.14
D(+)-biotiini ≥98%
D(+)-biotiini

Purity: 98+% CAS No.: 58-85-5 Molecular Formula: C10H16N2O3S Formula Weight: 244.32 Storage Temperature: 2°C to 8°C Melting Point: 224-226° Beilstein Reference: 86838 Merck Reference: 14,1231 Optical Rotation: +91° (c=1 in 0.1N NaOH) MDL No.: MFCD00005541


Essential vitamin that is important for amino acid and energy metabolism

Kaava: C₁₀H₁₆N₂O₃S
Sulamispiste: 224…226 °C
Tiheys: 1,41 g/cm³ (20 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00005541
CAS-numero: 58-85-5
EINECS: 200-399-3
Merck Index: 14,1231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR