2-(Trifluorimetyyli)bentseenisulfonyylikloridi ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A14108.03 A14108.06 A14108.14 A14108.22
A14108.03EA 57.2 EUR
A14108.03 A14108.06 A14108.14 A14108.22
2-(Trifluorimetyyli)bentseenisulfonyylikloridi ≥98%
2-(Trifluorimetyyli)bentseenisulfonyylikloridi

Purity: 98% CAS No.: 776-04-5 Molecular Formula: C7H4ClF3O2S Formula Weight: 244.62 Flash Point: 110°(230°F) Refractive Index: 1.5030 Melting Point: 22-24° Boiling Point: 133-135°/14mm Beilstein Reference: 2651854 UN No.: 3261 MDL No.: MFCD00051696

Kaava: C₇H₄ClF₃O₂S
Kiehumispiste: 133…135 °C (14 mmHg)
Sulamispiste: 22…24 °C
Tiheys: 1,585
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051696
CAS-numero: 776-04-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR