1,3,5-Trimetoksibentseeni ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

A13981.09 A13981.18 A13981.30
A13981.09EA 63.6 EUR
A13981.09 A13981.18 A13981.30
1,3,5-Trimetoksibentseeni ≥99%
1,3,5-Trimetoksibentseeni

Purity: 99% CAS No.: 621-23-8 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 85°(185°F) Melting Point: 51-53° Boiling Point: 255° Beilstein Reference: 1307993 MDL No.: MFCD00008385

Kaava: C₉H₁₂O₃
Kiehumispiste: 255 °C
Sulamispiste: 51…53 °C
Leimahduspiste: 85 °C (185 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008385
CAS-numero: 621-23-8
EINECS: 210-673-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR