4'-Fluoriasetofenoni ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A13950.14 A13950.22 A13950.36
A13950.14EA 69.1 EUR
A13950.14 A13950.22 A13950.36
4'-Fluoriasetofenoni ≥99%
4'-Fluoriasetofenoni

Purity: 99% CAS No.: 403-42-9 Molecular Formula: C8H7FO Formula Weight: 138.14 Flash Point: 71°(159°F) Refractive Index: 1.5110 Melting Point: 4° Boiling Point: 194-196° Beilstein Reference: 386013 MDL No.: MFCD00000354

Kaava: C₈H₇FO
Kiehumispiste: 194…196 °C
Sulamispiste: 4 °C
Tiheys: 1,141
Leimahduspiste: 71 °C (159 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000354
CAS-numero: 403-42-9
EINECS: 206-960-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR