Ethyl acetopyruvate ≥98%

Toimittaja: Thermo Fisher Scientific

A13808.14 A13808.06 A13808.22
A13808.14EA 401 EUR
A13808.14 A13808.06 A13808.22
Ethyl acetopyruvate ≥98%
Ethyl acetopyruvate

Purity: 98% CAS No.: 615-79-2 Molecular Formula: C7H10O4 Formula Weight: 158.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4730 Melting Point: 16-18° Boiling Point: 108-109°/12mm Beilstein Reference: 607062 MDL No.: MFCD00009124

Kaava: C₇H₁₀O₄
Kiehumispiste: 108…109 °C (12 mmHg)
Sulamispiste: 16…18 °C
Tiheys: 1,126
Leimahduspiste: >110 °C (230 °F)
MDL numero: MFCD00009124
CAS-numero: 615-79-2
EINECS: 210-447-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR