6-Metyyli-2,4-heptaanidioni ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A13373.04 A13373.09 A13373.18
A13373.04EA 56.5 EUR
A13373.04 A13373.09 A13373.18
6-Metyyli-2,4-heptaanidioni ≥98%
6-Metyyli-2,4-heptaanidioni

Purity: 98+% CAS No.: 3002-23-1 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 58°(136°F) Refractive Index: 1.4560 Boiling Point: 183° Beilstein Reference: 1751122 UN No.: 1224 MDL No.: MFCD00008939

Kaava: C₈H₁₄O₂
Kiehumispiste: 183 °C
Tiheys: 0,919
Leimahduspiste: 58 °C (136 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008939
CAS-numero: 3002-23-1
EINECS: 221-093-6
UN: 1224
ADR: 3,III

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR