4-Bromothiophene-2-carbaldehyde 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

A13304.06 A13304.22 A13304.14 AB114314.0100
A13304.06EA 65.7 EUR
A13304.06 A13304.22 A13304.14 ABCRAB114314.0100
4-Bromothiophene-2-carbaldehyde 96%
4-Bromothiophene-2-carbaldehyde

Purity: 96% CAS No.: 18791-75-8 Molecular Formula: C5H3BrOS Formula Weight: 191.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 114-115°/11mm MDL No.: MFCD00005431

Kaava: C₅H₃BrOS
Kiehumispiste: 114…115 °C (11 mmHg)
Sulamispiste: 44…48 °C
Leimahduspiste: >110 °C (230 °F)
MDL numero: MFCD00005431
CAS-numero: 18791-75-8
EINECS: 242-577-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR