Muurahaishappo 97%

Toimittaja: Thermo Fisher Scientific
Vaara

A13285.0C A13285.0E A13285.36
A13285.0CEA 394 EUR
A13285.0C A13285.0E A13285.36
Muurahaishappo 97%
Muurahaishappo

Purity: 97% CAS No.: 64-18-6 Molecular Formula: CH2O2 Formula Weight: 46.03 Flash Point: 50°(156°F) Refractive Index: 1.3710 Melting Point: 7-9° Boiling Point: 100-101° Beilstein Reference: 1209246 Merck Reference: 14,4241 UN No.: 1779 MDL No.: MFCD00003297

Kaava: HCO₂H
Kiehumispiste: 100…101 °C
Sulamispiste: 7…9 °C
Tiheys: 1,220
Leimahduspiste: 50 °C (156 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003297
CAS-numero: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
Merck Index: 14,4241
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR