Natrium-L(+)-glutamaatti-H2O ≥98%

Toimittaja: Thermo Fisher Scientific

A12919.0B A12919.0I A12919.30
A12919.0BEA 146 EUR
A12919.0B A12919.0I A12919.30
Natrium-L(+)-glutamaatti-H2O ≥98%
Natrium-L(+)-glutamaatti-H2O

Purity: 98+% CAS No.: 6106-04-3 Molecular Formula: C5H8NNaO4.H2O Formula Weight: 187.14 (169.12anhy) Melting Point: 228° dec. Beilstein Reference: 4164348

Kaava: C₅H₈NNaO₄·H₂O
Leimahduspiste: >225 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150138
CAS-numero: 6106-04-3
EINECS: 205-538-1
Merck Index: 13,06278

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR