1,1,1,3,3,3-Heksafluori-2-propanoli ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

A12747.06 A12747.14 A12747.22 A12747.99
A12747.06EA 84.3 EUR
A12747.06 A12747.14 A12747.22 A12747.99
1,1,1,3,3,3-Heksafluori-2-propanoli ≥99%
1,1,1,3,3,3-Heksafluori-2-propanoli

Purity: 99+% CAS No.: 920-66-1 Molecular Formula: C3H2F6O Formula Weight: 168.04 Refractive Index: 1.2750 Melting Point: -4° Boiling Point: 59-60° Beilstein Reference: 1841007 UN No.: 1760 MDL No.: MFCD00011651

Kaava: C₃H₂F₆O
Kiehumispiste: 59…60 °C
Sulamispiste: –4 °C
Tiheys: 1,619
Leimahduspiste: >100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011651
CAS-numero: 920-66-1
EINECS: 213-059-4
UN: 1760
ADR: 8,II
Merck Index: 13,09598

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR