Methyl-p-anisate ≥98%

Toimittaja: Thermo Fisher Scientific

A12598.14 A12598.22
A12598.14EA 90.9 EUR
A12598.14 A12598.22
Methyl-p-anisate ≥98%
Methyl-p-anisate

Purity: 98+% CAS No.: 121-98-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Odour: Characteristic Boiling Point: 244-245° Beilstein Reference: 2208571 MDL No.: MFCD00008437

Kaava: C₉H₁₀O₃
Kiehumispiste: 244…245 °C
Sulamispiste: 48…51 °C
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008437
CAS-numero: 121-98-2
EINECS: 204-513-2

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR