1-Fluori-2-jodibentseeni ≥99% stabiloitu

Toimittaja: Thermo Fisher Scientific
Varoitus


Stabilization: Stabilised with copper

A12411.14 A12411.22 A12411.36
A12411.14EA 109 EUR
A12411.14 A12411.22 A12411.36
1-Fluori-2-jodibentseeni ≥99% stabiloitu
1-Fluori-2-jodibentseeni

Stabilized with copper


Purity: 99% CAS No.: 348-52-7 Molecular Formula: C6H4FI Formula Weight: 222.00 Flash Point: 71°(159°F) Refractive Index: 1.5900 Melting Point: -43° to -40° Boiling Point: 188-189° Beilstein Reference: 2039770 MDL No.: MFCD00001031

Kaava: C₆H₄FI
Kiehumispiste: 188…189 °C
Sulamispiste: –43 °C to –40 °C
Tiheys: 1,903
Leimahduspiste: 71 °C (159 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001031
CAS-numero: 348-52-7
EINECS: 206-477-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR