Pyridiini-2,6-dikarboksyylihappo ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A12263.14 A12263.22 A12263.36
A12263.14EA 59.1 EUR
A12263.14 A12263.22 A12263.36
Pyridiini-2,6-dikarboksyylihappo ≥98%
Pyridiini-2,6-dikarboksyylihappo

Purity: 98% CAS No.: 499-83-2 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Flash Point: 188°(370°F) Melting Point: ca 245° dec. Beilstein Reference: 131629 MDL No.: MFCD00006299

Kaava: C₇H₅NO₄
Kiehumispiste: 357 °C (1013 hPa)
Leimahduspiste: 188 °C (370 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006299
CAS-numero: 499-83-2
EINECS: 207-894-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR