4-tert-Butyylitiofenoli ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

A12148.03 A12148.06 A12148.14
A12148.03EA 65.3 EUR
A12148.03 A12148.06 A12148.14
4-tert-Butyylitiofenoli ≥97%
4-tert-Butyylitiofenoli

Purity: 97% CAS No.: 2396-68-1 Molecular Formula: C10H14S Formula Weight: 166.29 Flash Point: 110°(230°F) Refractive Index: 1.5485 Odour: Stench Melting Point: -11° Boiling Point: 238° Beilstein Reference: 606476 MDL No.: MFCD00022067

Kaava: C₁₀H₁₄S
Kiehumispiste: 238 °C
Sulamispiste: –11 °C
Tiheys: 0,990
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00022067
CAS-numero: 2396-68-1
EINECS: 219-255-6
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR