Calcium bis(dihydrogenorthophosphate) hydrate ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

A12123.0C A12123.0E A12123.36
A12123.0CEA 1120 EUR
A12123.0C A12123.0E A12123.36
Calcium bis(dihydrogenorthophosphate) hydrate ≥97%
Calcium bis(dihydrogenorthophosphate) hydrate

Purity: 97% CAS No.: 301524-28-7 Molecular Formula: Ca(H2PO4)2.xH2O Formula Weight: 234.06(anhy) Flash Point: None Melting Point: ca 200° dec. MDL No.: MFCD00010898

Kaava: H₄CaO₈P₂·H₂O
Sulamispiste: ∼ 200 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010898
CAS-numero: 301524-28-7
EINECS: 231-837-1
Merck Index: 14,1693

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR