1-Bromi-4-(trifluorimetoksi)bentseeni ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A12104.06 A12104.14
A12104.06EA 112 EUR
A12104.06 A12104.14
1-Bromi-4-(trifluorimetoksi)bentseeni ≥98%
1-Bromi-4-(trifluorimetoksi)bentseeni

Purity: 98% CAS No.: 407-14-7 Molecular Formula: C7H4BrF3O Formula Weight: 241.01 Flash Point: 67°(152°F) Refractive Index: 1.4610 Odour: Unpleasant Boiling Point: 155-157° Beilstein Reference: 2046332 MDL No.: MFCD00040834

Kaava: C₇H₄BrF₃O
Kiehumispiste: 155…157 °C
Tiheys: 1,622
Leimahduspiste: 67 °C (152 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00040834
CAS-numero: 407-14-7
EINECS: 206-979-2
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR