2',4'-Dihydroksiasetofenoni ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A12066.18 A12066.30
A12066.18EA 93.5 EUR
A12066.18 A12066.30
2',4'-Dihydroksiasetofenoni ≥98%
2',4'-Dihydroksiasetofenoni

Purity: 98% CAS No.: 89-84-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 142-146° Beilstein Reference: 1282505 Merck Reference: 14,8140 MDL No.: MFCD00002279

Kaava: C₈H₈O₃
Kiehumispiste: 336 °C (1013 hPa)
Sulamispiste: 142…146 °C
Tiheys: 1,18 g/cm³ (141 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002279
CAS-numero: 89-84-9
EINECS: 201-945-3
Merck Index: 14,8140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR