3,4,5-Tribromo-1H-pyrazole ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

A12031.06 A12031.14
A12031.06EA 82.1 EUR
A12031.06 A12031.14
3,4,5-Tribromo-1H-pyrazole ≥97%
3,4,5-Tribromo-1H-pyrazole

Purity: 97% CAS No.: 17635-44-8 Molecular Formula: C3HBr3N2 Formula Weight: 304.78 Odour: Odourless Melting Point: 184-188° Beilstein Reference: 113501 MDL No.: MFCD00040248

Kaava: C₃HBr₃N₂
Sulamispiste: 184…188 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00040248
CAS-numero: 17635-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR