(4-Carboxybutyl)triphenylphosphonium bromide ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A12023.14 A12023.22 A12023.36
A12023.14EA 110 EUR
A12023.14 A12023.22 A12023.36
(4-Carboxybutyl)triphenylphosphonium bromide ≥98%
(4-Carboxybutyl)triphenylphosphonium bromide

Purity: 98% CAS No.: 17814-85-6 Molecular Formula: C23H24BrO2P Formula Weight: 443.33 Flash Point: 195°(383°F) Melting Point: 205-209° Beilstein Reference: 3586477 MDL No.: MFCD00011906

Kaava: C₂₃H₂₄BrO₂P
Sulamispiste: 205…209 °C
Leimahduspiste: 195 °C (383 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011906
CAS-numero: 17814-85-6
EINECS: 241-782-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR