Pyridinium chlorochromate ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A11752.14 A11752.22 A11752.36 A11752.PG
A11752.14EA 55.5 EUR
A11752.14 A11752.22 A11752.36 A11752.PG
Pyridinium chlorochromate ≥98%
Pyridinium chlorochromate

Purity: 98% CAS No.: 26299-14-9 Molecular Formula: C5H6ClCrNO3 Formula Weight: 215.56 Melting Point: ca 206° dec. Merck Reference: 14,7974 MDL No.: MFCD00013106

Kaava: C₅H₆ClCrNO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013106
CAS-numero: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 14,7974

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR