Kaliumkloridi ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A11662.0B A11662.0G A11662.0I
A11662.0BEA 95.2 EUR
A11662.0B A11662.0G A11662.0I
Kaliumkloridi ≥99%
Kaliumkloridi

Purity: 99% CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Refractive Index: 1.490 Odour: Odourless Boiling Point: 1500° subl. Merck Reference: 14,7621 MDL No.: MFCD00011360

Kaava: KCl
Kiehumispiste: 1500 °C (subl.)
Sulamispiste: 773 °C
Tiheys: 1,984
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011360
CAS-numero: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,7621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR