Fluoreseiini, dinatriumsuola hydraatti

Toimittaja: Thermo Fisher Scientific

A11659.0E A11659.22 A11659.36
A11659.0EEA 788 EUR
A11659.0E A11659.22 A11659.36
Fluoreseiini, dinatriumsuola hydraatti
Fluoreseiini, dinatriumsuola hydraatti

Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.


CAS No.: 518-47-8 Molecular Formula: C20H10Na2O5.xH2O Formula Weight: 376.28 Odour: Odourless pH: 8.3 Melting Point: 250° Beilstein Reference: 3833041 Merck Reference: 14,4159 MDL No.: MFCD00167039

Kaava: C₂₀H₁₀Na₂O₅·xH₂O
Sulamispiste: >250 °C
Tiheys: 1,601 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00167039
CAS-numero: 518-47-8
EINECS: 208-253-0
Merck Index: 14,4159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR