Litiumfluoridi 97%

Toimittaja: Thermo Fisher Scientific
Vaara

A11632.0E A11632.22 A11632.36
A11632.0EEA 2090 EUR
A11632.0E A11632.22 A11632.36
Litiumfluoridi 97%
Litiumfluoridi

Purity: 98+% CAS No.: 7789-24-4 Molecular Formula: LiF Formula Weight: 25.94 Refractive Index: 1.3915 Odour: Odourless Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090

Kaava: FLi
Kiehumispiste: 1680 °C
Sulamispiste: 845 °C
Tiheys: 2,635
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011090
CAS-numero: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5531

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR