Sodium dodecatungstophosphate hydrate

Toimittaja: Thermo Fisher Scientific
Varoitus

A11575.14 A11575.22 A11575.36
A11575.14EA 139 EUR
A11575.14 A11575.22 A11575.36
Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate

CAS No.: 51312-42-6 Molecular Formula: Na3PO4.12WO3.xH2O Formula Weight: 2946.12(anhy) Melting Point: 350° MDL No.: MFCD00151658

Kaava: Na₃O₄₀PW₁₂·H₂O
Sulamispiste: 350 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151658
CAS-numero: 51312-42-6
EINECS: 257-132-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR