Bis(cyclopentadienyl)titanium dichloride ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

A11456.09 A11456.18 A11456.30
A11456.09EA 102 EUR
A11456.09 A11456.18 A11456.30
Bis(cyclopentadienyl)titanium dichloride ≥97%
Bis(cyclopentadienyl)titanium dichloride

Purity: 97% CAS No.: 1271-19-8 Molecular Formula: C10H10Cl2Ti Formula Weight: 249.00 Odour: Odourless Melting Point: ca 285° dec. MDL No.: MFCD00003723

Kaava: C₁₀H₁₀Cl₂Ti
Sulamispiste: ∼ 285 °C (haj. lämpötila)
Tiheys: 1,60
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003723
CAS-numero: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II
Merck Index: 14,9482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR