4-Tolualdehydi ≥98%
Toimittaja: Thermo Fisher Scientific
A11384.22 A11384.06 A11384.0E A11384.36
A11384.22EA 72.4 EUR
A11384.22 A11384.06 A11384.0E A11384.36
4-Tolualdehydi ≥98%
4-Tolualdehydi
Purity: 98% CAS No.: 104-87-0 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 80°(176°F) Refractive Index: 1.5460 Melting Point: -6° Boiling Point: 204° Beilstein Reference: 385772 MDL No.: MFCD00006954