2-Methyl-4-pentenoic acid ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A11297.09 A11297.18
A11297.09EA 108 EUR
A11297.09 A11297.18
2-Methyl-4-pentenoic acid ≥98%
2-Methyl-4-pentenoic acid

Purity: 98% CAS No.: 1575-74-2 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 107°(224°F) Refractive Index: 1.4310 Odour: Characteristic Boiling Point: 195° Beilstein Reference: 1720987 UN No.: 3265 MDL No.: MFCD00015570

Kaava: C₆H₁₀O₂
Kiehumispiste: 195 °C
Tiheys: 0,950
Leimahduspiste: 107 °C (224 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015570
CAS-numero: 1575-74-2
EINECS: 216-404-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR