2-Phenoxynicotinic acid ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A11247.03 A11247.06 A11247.14
A11247.03EA 52.5 EUR
A11247.03 A11247.06 A11247.14
2-Phenoxynicotinic acid ≥98%
2-Phenoxynicotinic acid

Purity: 98% CAS No.: 35620-71-4 Molecular Formula: C12H9NO3 Formula Weight: 215.21 Melting Point: 179-182° Beilstein Reference: 172906

Kaava: C₁₂H₉NO₃
Sulamispiste: 179…182 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014629
CAS-numero: 35620-71-4
EINECS: 252-647-5

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR