1-Heksadekanoli ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A11180.0E A11180.36
A11180.0EEA 227 EUR
A11180.0E A11180.36
1-Heksadekanoli ≥98%
1-Heksadekanoli

Purity: 98% CAS No.: 36653-82-4 Molecular Formula: CH3(CH2)15OH Formula Weight: 242.45 Flash Point: 135°(275°F) Physical Form: Crystalline Density: 0.817 Melting Point: 47-51° Boiling Point: 190°/15mm Solubility: Soluble in alcohol, chloroform, ether Beilstein Reference: 1748475 Merck Reference: 14,2028 MDL No.: MFCD00004760

Kaava: H₃C(CH₂)₁₄CH₂OH
Kiehumispiste: 190 °C (15 mmHg)
Sulamispiste: 47…51 °C
Tiheys: 0,817
Leimahduspiste: 135 °C (275 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004760
CAS-numero: 36653-82-4
EINECS: 253-149-0
Merck Index: 14,2028

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR