Hemin chloride (from porcine) 97+%

Toimittaja: Thermo Fisher Scientific

A11165.06 A11165.03 A11165.14
A11165.06EA 333 EUR
A11165.06 A11165.03 A11165.14
Hemin chloride (from porcine) 97+%
Hemin chloride

Stimulates synthesis of globulin.


Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.


Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water


Varoitus: Incompatible with strong oxidizing agents.

Kaava: C₃₄H₃₂ClFeN₄O₄
Sulamispiste: >300 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00010726
CAS-numero: 16009-13-5
EINECS: 240-140-1
Merck Index: 14,4644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR