(4-Fluorifenyyli)etikkahappo ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A10646.06 A10646.14 A10646.22
A10646.06EA 50.6 EUR
A10646.06 A10646.14 A10646.22
(4-Fluorifenyyli)etikkahappo ≥98%
(4-Fluorifenyyli)etikkahappo

Purity: 98% CAS No.: 405-50-5 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 100°(212°F) Odour: Odourless Boiling Point: 164°/2mm Beilstein Reference: 972145 Merck Reference: 14,4177 MDL No.: MFCD00004343

Kaava: (C₆H₄F)CH₂COOH
Kiehumispiste: 164 °C (2 mmHg)
Sulamispiste: 81…85 °C
Leimahduspiste: >100 °C (212 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004343
CAS-numero: 405-50-5
EINECS: 206-972-4
Merck Index: 14,4177

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR