DL-Threonine ≥99%

Toimittaja: Thermo Fisher Scientific

A10606.14 A10606.22 A10606.36
A10606.14EA 98.7 EUR
A10606.14 A10606.22 A10606.36
DL-Threonine ≥99%
DL-Threonine

Purity: 99% CAS No.: 80-68-2 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: ca 244° dec. Beilstein Reference: 1721647 Merck Reference: 14,9380 MDL No.: MFCD00063722

Kaava: C₄H₉NO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00063722
CAS-numero: 80-68-2
EINECS: 201-300-6
Merck Index: 14,9380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR