Vismutti(III)kloridi, (max. 3% H₂O) ≥98% (dry weight)

Toimittaja: Thermo Fisher Scientific
Vaara

A10510.14 A10510.22 A10510.36
A10510.14EA 108 EUR
A10510.14 A10510.22 A10510.36
Vismutti(III)kloridi, (max. 3% H₂O) ≥98% (dry weight)
Vismutti(III)kloridi

Purity: 98% CAS No.: 7787-60-2 Molecular Formula: BiCl3 Formula Weight: 315.34 Odour: Irritating Melting Point: 230-232° Boiling Point: ca 430° subl. Merck Reference: 14,1262 UN No.: 3260 MDL No.: MFCD00003461

Kaava: BiCl₃
Kiehumispiste: ∼430 °C (subl.)
Sulamispiste: 230…232 °C
Tiheys: 4,75
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003461
CAS-numero: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 14,1262

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR