2-Nitro-1,3,5-trimethoxybenzene ≥98%

Toimittaja: Thermo Fisher Scientific

A10473.06 A10473.14 A10473.22
A10473.06EA 179 EUR
A10473.06 A10473.14 A10473.22
2-Nitro-1,3,5-trimethoxybenzene ≥98%
2-Nitro-1,3,5-trimethoxybenzene

Purity: 98% CAS No.: 14227-18-0 Molecular Formula: C9H11NO5 Formula Weight: 213.19 Melting Point: 150-153° Beilstein Reference: 2277262

Kaava: C₉H₁₁NO₅
Sulamispiste: 150…153 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016992
CAS-numero: 14227-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR