Methyl-6-chloronicotinate ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A10396.06 A10396.14
A10396.06EA 115 EUR
A10396.06 A10396.14
Methyl-6-chloronicotinate ≥98%
Methyl-6-chloronicotinate

Purity: 98% CAS No.: 73781-91-6 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 83-88° Beilstein Reference: 128648

Kaava: C₇H₆ClNO₂
Sulamispiste: 83…88 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00023420
CAS-numero: 73781-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR