Sitruunahappo ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

A10395.0B A10395.0I A10395.30
A10395.0BEA 136 EUR
A10395.0B A10395.0I A10395.30
Sitruunahappo ≥99%
Sitruunahappo

Purity: 99+% CAS No.: 77-92-9 Molecular Formula: C6H8O7 Formula Weight: 192.13 Odour: Odourless pH: 1,6 Melting Point: 153-154° Beilstein Reference: 782061 Merck Reference: 14,2326 MDL No.: MFCD00011669

Kaava: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Kiehumispiste: 310
Sulamispiste: 153…154 °C
Tiheys: 1.665
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011669
CAS-numero: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR