tert-Butyl-N-hydroxycarbamate ≥98%

Toimittaja: Thermo Fisher Scientific

A10349.03 A10349.06 A10349.14
A10349.03EA 70.5 EUR
A10349.03 A10349.06 A10349.14
tert-Butyl-N-hydroxycarbamate ≥98%
tert-Butyl-N-hydroxycarbamate

Purity: 98+% CAS No.: 36016-38-3 Molecular Formula: (CH3)3COCONHOH Formula Weight: 133.15 Storage Temperature: 2°C to 8°C Melting Point: 55-58° Beilstein Reference: 1756546 MDL No.: MFCD00002107

Kaava: C₅H₁₁NO₃
Sulamispiste: 55…58 °C
MDL numero: MFCD00002107
CAS-numero: 36016-38-3
EINECS: 252-836-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR