3,3-Dimetyylibutanoni ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone

A10289.22 A10289.36
A10289.22EA 217 EUR
A10289.22 A10289.36
3,3-Dimetyylibutanoni ≥97%
3,3-Dimetyylibutanoni

Purity: 97% CAS No.: 75-97-8 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 5°(41°F) Refractive Index: 1.3970 Melting Point: -53° Boiling Point: 105-106° Merck Reference: 14,7440 UN No.: 1224 MDL No.: MFCD00008846

Kaava: C₆H₁₂O
Kiehumispiste: 105…106 °C
Sulamispiste: –53 °C
Tiheys: 0,803
Leimahduspiste: 5 °C (41 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008846
CAS-numero: 75-97-8
EINECS: 200-920-4
UN: 1224
ADR: 3,II
Merck Index: 14,7440

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR