3-Bromobutanone ≥97% stabiloitu

Toimittaja: Thermo Fisher Scientific
Vaara


Stabilization: Stabiised with magnesium oxide (MgO) < 1 %

A10186.30 A10186.18 A10186.09
A10186.30EA 2510 EUR
A10186.30 A10186.18 A10186.09
3-Bromobutanone ≥97% stabiloitu
3-Bromobutanone

Stabilized with 1% magnesium oxide


Purity: 97% CAS No.: 814-75-5 Molecular Formula: C4H7BrO Formula Weight: 151.01 Flash Point: 100°(212°F) Refractive Index: 1.4590 Odour: Pungent Boiling Point: 69-70°/70mm Beilstein Reference: 906773 UN No.: 2922 MDL No.: MFCD00013538

Kaava: H₃CC(O)BrCHCH₃
Kiehumispiste: 69…70 °C (70 mmHg)
Tiheys: 1,438
Leimahduspiste: >100 °C (212 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013538
CAS-numero: 814-75-5
EINECS: 212-404-6
UN: 2922
ADR: 8,II

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR