(4-Etoksifenyyli)etikkahappo ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A10129.09 A10129.14 A10129.18
A10129.09EA 81.6 EUR
A10129.09 A10129.14 A10129.18
(4-Etoksifenyyli)etikkahappo ≥98%
(4-Etoksifenyyli)etikkahappo

Purity: 98+% CAS No.: 4919-33-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 87-91° Beilstein Reference: 1103143

Kaava: (C₆H₄(OCH₂CH₃))CH₂COOH
Kiehumispiste: 313 °C (1013 hPa)
Sulamispiste: 87…91 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004346
CAS-numero: 4919-33-9
EINECS: 225-545-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR