VWR®, Withdrawal System for 30 L Stainless Steel Barrels, Vertical
Toimittaja: VWR Chemicals
99907.030EA 578 EUR
99907.030
VWR®, Withdrawal System for 30 L Stainless Steel Barrels, Vertical
Pumput Rumpupumput
Specially developed solvent withdrawal system for inert gas pressurizing.
  • Withdrawal system for 30 L barrels (stainless steel)
  • Perfluorelastomer o-rings
  • Includes 1 dip tube for 30 L barrels
Specially developed solvent withdrawal system for inert gas pressurizing for VWR® returnable 30 L stainless steel barrels.
Lisävarusteiden tiedot: Ideal for use with flexible hose with filling nozzle (99908.001).