VWR®, Density standards, Certified Reference Materials, referenced at 15 °C

Toimittaja: VWR Chemicals

99624.600EA 85.7 EUR
99624.600 99626.600 99628.600 99630.600 99634.600
VWR®, Density standards, Certified Reference Materials, referenced at 15 °C
Standardiaineet ja vertailumateriaalit Density Standards

VWR® Chemicals ISO 17025/ISO 17034 dual certified-density standards are for the verification of instruments used to measure the density of materials which are fluid at the desired test temperature.


  • ISO 17025 / ISO 17034 certified under UKAS accreditation
  • Density tested in strict accordance with ASTM D1480, a primary method for density determination
  • Density standards range between 0,6654 to 1,6300 g/ml
  • Ensures full compliance to ASTM and IP test method protocols
  • Shelf life between 12/24 months

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR