VWR®, General purpose viscosity standards

Toimittaja: VWR Chemicals

99438.260EA 91.6 EUR
99438.260 99439.260 99440.260 99441.260 99442.260 99443.260 99444.260 99445.260 99446.260 99447.260 99448.260 99449.260 99450.260 99451.260 99452.260 99453.260 99454.260 99455.260 99456.260 99457.260 99458.260 99459.260 99460.260 99461.260 99462.260 99463.260 99464.260 99465.260 99466.260 99467.260 99468.260 99469.260 99470.260 99471.260 99472.260 99473.260 99474.260 99475.260 99476.260 99477.260
VWR®, General purpose viscosity standards
Standardiaineet ja vertailumateriaalit Viskometriastandardit

These ISO 17025 and ISO 17034 certified general purpose viscosity standards provide certified kinematic viscosity, dynamic viscosity and density data across temperature ranges from 20 to 100 °C.


  • Certified in strict accordance with ASTM D2162
  • Density g/ml in accordance with ASTM D1480
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) and dynamic viscosity (mPa·s) given at all temperatures
  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging


Certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture.


These products have been tested at primary level using master viscometers in accordance with ASTM D2162 to ensure the lowest uncertainty of measurement, thus providing standards that are certified with primary level accuracy. ASTM D2162 is the only referenced method for the certification of viscosity standards when adopting ASTM test method protocols.


Sertifikaatit: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.


Varoitus: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR