VWR®, Total Base Number (TBN) standards
Toimittaja: VWR Chemicals
99352.202EA 258 EUR
99352.202 99352.001 99353.154 99353.001 99354.154 99354.001 99355.154 99355.001 99356.154 99356.001 99357.154 99357.001 99358.154 99358.001 99359.154 99359.001
VWR®, Total Base Number (TBN) standards
Standardiaineet ja vertailumateriaalit Petroleum Industry Standards
Total Base Number (TBN) standards are specifically manufactured for the verification of analytical instruments used to determine base number by potentiometric titration. Each standard is certified in strict accordance with international test method protocols, ASTM D2896 / IP 276.
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Base Number (BN) tested and certified in accordance with ASTM D2896 / IP 276
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
  • Supplied in tamper-evident glass packaging, providing assurance of sample integrity
Sertifikaatit: Manufacture and certification is carried out in strict accordance with dual-UKAS accreditations to ISO 17025 and ISO 17034. This dual-accreditation ensures the highest level of accreditation guarantee, and provides the most credible certified data available worldwide.
Varoitus: The concentration value given in the ordering table below is the nominal concentration for the TBN value - actual batch values are given in the certification documents supplied with the product and also on the label.