VWR®, Certified Reference Material, Pensky Martens flash point
Toimittaja: VWR Chemicals
99112.230EA 292 EUR
99112.230 99113.230 99115.230 99152.001 99153.001 99154.001
VWR®, Certified Reference Material, Pensky Martens flash point
Standardiaineet ja vertailumateriaalit Petroleum Industry Standards
Flash point Certified Reference Materials are for the verification of laboratory flash point instruments that are used in testing petroleum and derivative products. PMCC flash point standards are tested and certified in accordance with ASTM D93 'Standard Test Method for Flash Point by Pensky Martens Closed Cup Tester - Procedure A'
  • Fully certified in accordance with ISO 17025 and ISO 17034
  • Highest level of accreditation guaranteed, providing the most credible data currently available worldwide
  • All data generated exclusively by ISO 17025 accredited independent laboratories
  • Ensures good inter-laboratory correlation
  • Certified in accordance with ASTM / EN or IP Test method protocols
  • Fully traceable to international standards
  • Supplied in tamper-evident UN compliant security packs
These CRMs are used for Pensky-Martens Closed Cup (ASTM D93, IP 34 and ISO 2719).
Sertifikaatit: UKAS ISO 17025 and ISO 17034
Varoitus: The flash point given in the ordering table below is the nominal flash point - actual batch values are given in the certification documents supplied with the product and also on the label.