J.T.Baker®, Female QT-2 Quick Connect Bodies
Toimittaja: Avantor
9802-01EA 0 EUR
9802-01 9802-02 9802-03 9801-00 9802-04 9802-05 9802-06 9802-07 9802-00
J.T.Baker®, Female QT-2 Quick Connect Bodies
Kiinnitykset ja liittimet
Swagelock colour-coded quick connect for use with various solvents and gas ports.
  • For use with female tube fittings
  • For less than 1250 L sizes