1-Propanoli, Supelco®

Toimittaja: Merck
Vaara

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

M-Clarity™ quality level = MQ100

96566-10ML-F 96566-5ML-F
96566-10ML-FEA 277 EUR
96566-10ML-F 96566-5ML-F
1-Propanoli, Supelco®
1-Propanoli
Kaava: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Kiehumispiste: 97
Sulamispiste: –127 °C
Tiheys: 0.8053 g/cm³ (20 °C)
Leimahduspiste: 15
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002941
CAS-numero: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce4d0cad53f180e9772a3503dfcde3eacf5c5940&vid=aa6e7c47873264a837a356f6c99c07a1af5c4e09&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR