Typpihappo 69,0 - 70,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Nitric acid solution

9598-51EA 128.72 EUR
9598-51 9598-34 9598-00
Typpihappo 69,0 - 70,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Typpihappo

For trace metal analysis. Lot analysis on label.

Kaava: HNO₃
MW: 63.01 g/mol
Kiehumispiste: 87
Sulamispiste: –42 °C
Tiheys: 1.51
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011349
CAS-numero: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06608

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use 69.0 - 70.0 %
ACS - Assay (HNO₃) 69.0 - 70.0 %
Appearance Passes Test
ACS - Color (APHA) ≤ 10
ACS - Residue after Ignition ≤ 2 ppm
Chloride (Cl) ≤ 0.04 ppm
Phosphate (PO₄) ≤ 0.10 ppm
Sulfate (SO₄) ≤ 0.4 ppm
Trace Impurities - Aluminum (Al) ≤ 30.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 1.0 ppb
Trace Impurities - Beryllium (Be) ≤ 1.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 1.0 ppb
Trace Impurities - Boron (B) ≤ 4.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 1.0 ppb
Trace Impurities - Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Chromium (Cr) ≤ 10.0 ppb
Trace Impurities - Cobalt (Co) ≤ 1.0 ppb
Trace Impurities - Copper (Cu) ≤ 1.0 ppb
Trace Impurities - Gallium (Ga) ≤ 20.0 ppb
Trace Impurities - Germanium (Ge) ≤ 4.0 ppb
Trace Impurities - Gold (Au) ≤ 4.0 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 10.0 ppb
Trace Impurities - Lead (Pb) ≤ 0.5 ppb
Trace Impurities - Lithium (Li) ≤ 1.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 7.0 ppb
Trace Impurities - Manganese (Mn) ≤ 1.0 ppb
Trace Impurities - Mercury (Hg) ≤ 0.5 ppb
Trace Impurities - Molybdenum (Mo) ≤ 5.0 ppb
Trace Impurities - Nickel (Ni) ≤ 1.0 ppb
Trace Impurities - Niobium (Nb) ≤ 1.0 ppb
Trace Impurities - Potassium (K) ≤ 5.0 ppb
Trace Impurities - Silicon (Si) ≤ 20.0 ppb
Trace Impurities - Silver (Ag) ≤ 1.0 ppb
Trace Impurities - Sodium (Na) ≤ 200.0 ppb
Trace Impurities - Strontium (Sr) ≤ 1.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 2.0 ppb
Trace Impurities - Thallium (Tl) ≤ 5.0 ppb
Trace Impurities - Tin (Sn) ≤ 5.0 ppb
Trace Impurities - Vanadium (V) ≤ 1.0 ppb
Trace Impurities - Zinc (Zn) ≤ 5.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 1.0 ppb
Meets ACS Specifications


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR