Micro particles based on melamine resin, analytical standard, Supelco®
Toimittaja: Merck
91329-10ML-F 95523-5ML-F 96062-10ML-F
91329-10ML-FEA 403 EUR
91329-10ML-F 95523-5ML-F 96062-10ML-F
Micro particles based on melamine resin, analytical standard, Supelco®
Standardiaineet ja vertailumateriaalit Partikkelikokostandardit
For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acccad2f6f908f63022098359687870eba2301cb&vid=acccad2f6f908f63022098359687870eba2301cb&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffb340ddacfd75078f6f41ed86e81b569f95fe29&vid=ffb340ddacfd75078f6f41ed86e81b569f95fe29&prid=web&clid=SAPDAM